COM448684414 Combi-Blocks, Inc.

Methyl 7-bromo-2-methyl-1-propyl-1,3-benzodiazole-5-carboxylate

  • MDL number MFCD24448848
  • Cas Number : 1437794-72-3
  • Molecular Weight : 311.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9224

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9224
Availability details for
Quantity
  • Availability
  • Add to Cart