COM448684415 Combi-Blocks, Inc.

5-Bromo-2-methanesulfonylphenol

  • MDL number MFCD23699518
  • Cas Number : 1426958-41-9
  • Molecular Weight : 251.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9228

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9228
Availability details for
Quantity
  • Availability
  • Add to Cart