COM448684425 Combi-Blocks, Inc.

7-Bromo-1-propyl-1,3-benzodiazole-5-carboxylic acid

  • MDL number MFCD24448849
  • Cas Number : 1437794-76-7
  • Molecular Weight : 283.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9252

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9252
Availability details for
Quantity
  • Availability
  • Add to Cart