COM448684427 Combi-Blocks, Inc.

7-Bromo-1-propyl-1,2,3-benzotriazole-5-carboxylic acid

  • MDL number MFCD26131169
  • Cas Number : 1820666-73-6
  • Molecular Weight : 284.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9254

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9254
Availability details for
Quantity
  • Availability
  • Add to Cart