COM448684430 Combi-Blocks, Inc.

Ethyl 1-ethyl-1,3-benzodiazole-5-carboxylate

  • MDL number MFCD25953927
  • Cas Number : 1354408-65-3
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9258

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9258
Availability details for
Quantity
  • Availability
  • Add to Cart