COM448684437 Combi-Blocks, Inc.

6-(Benzyloxy)-1-methyl-1,3-benzodiazole

  • MDL number MFCD25121772
  • Cas Number : 1445322-55-3
  • Molecular Weight : 238.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9272

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9272
Availability details for
Quantity
  • Availability
  • Add to Cart