COM448684441 Combi-Blocks, Inc.

2-Nitro-4-sulfamoylbenzoic acid

  • MDL number MFCD27981346
  • Cas Number : 29092-31-7
  • Molecular Weight : 246.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9277

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9277
Availability details for
Quantity
  • Availability
  • Add to Cart