COM448684476 Combi-Blocks, Inc.

Ethyl 4-(cyclopropylamino)-3-nitrobenzoate

  • MDL number MFCD14635824
  • Cas Number : 1467047-28-4
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9322

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9322
Availability details for
Quantity
  • Availability
  • Add to Cart