COM448684490 Combi-Blocks, Inc.

4-Amino-3-chloro-5-nitrobenzonitrile

  • MDL number MFCD25953942
  • Cas Number : 1456531-72-8
  • Molecular Weight : 197.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9363

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9363
Availability details for
Quantity
  • Availability
  • Add to Cart