COM448684510 Combi-Blocks, Inc.

4-Bromo-2-chloro-1-methanesulfonylbenzene

  • MDL number MFCD26131172
  • Cas Number : 648905-09-3
  • Molecular Weight : 269.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9394

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9394
Availability details for
Quantity
  • Availability
  • Add to Cart