COM448684513 Combi-Blocks, Inc.

5-(Trifluoromethyl)-1H-benzo[d]imidazole HCl

  • MDL number MFCD08460609
  • Cas Number : 1260642-93-0
  • Molecular Weight : 222.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9398

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9398
Availability details for
Quantity
  • Availability
  • Add to Cart