COM448684522 Combi-Blocks, Inc.

4-(4-Chlorophenyl)-1H-pyrazole

  • MDL number MFCD03647018
  • Cas Number : 111016-47-8
  • Molecular Weight : 178.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9413

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9413
Availability details for
Quantity
  • Availability
  • Add to Cart