COM448684527 Combi-Blocks, Inc.

7-Chloro-4,4-dimethyl-1,3-dihydroquinolin-2-one

  • MDL number MFCD11040449
  • Cas Number : 133999-06-1
  • Molecular Weight : 209.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9418

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9418
Availability details for
Quantity
  • Availability
  • Add to Cart