COM448684559 Combi-Blocks, Inc.

3-(1,2,4-Oxadiazol-3-yl)pyridine

  • MDL number MFCD00465052
  • Cas Number : 10550-16-0
  • Molecular Weight : 147.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9463

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9463
Availability details for
Quantity
  • Availability
  • Add to Cart