COM448684565 Combi-Blocks, Inc.

1,3-Dibromo-5-methanesulfonylbenzene

  • MDL number MFCD27936857
  • Cas Number : 1222407-24-0
  • Molecular Weight : 314.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9471

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9471
Availability details for
Quantity
  • Availability
  • Add to Cart