COM448684569 Combi-Blocks, Inc.

Methyl 3-chloro-4-nitrobenzoate

  • MDL number MFCD11977375
  • Cas Number : 243984-48-7
  • Molecular Weight : 215.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-chloro-4-nitrobenzoate
Inchi Code 1S/C8H6ClNO4/c1-14-8(11)5-2-3-7(10(12)13)6(9)4-5/h2-4H,1H3
Inchi Key UHPKNDYXGXJKMH-UHFFFAOYSA-N

Description

Purity 98%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9476

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9476
Availability details for
Quantity
  • Availability
  • Add to Cart