COM448684571 Combi-Blocks, Inc.

1-Bromo-4-methanesulfonyl-2-(trifluoromethyl)benzene

  • MDL number MFCD15527241
  • Cas Number : 1820711-75-8
  • Molecular Weight : 303.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9480

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9480
Availability details for
Quantity
  • Availability
  • Add to Cart