COM448684572 Combi-Blocks, Inc.

1-(methylsulfanyl)-3-nitrobenzene

  • MDL number MFCD00024501
  • Cas Number : 2524-76-7
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9481

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9481
Availability details for
Quantity
  • Availability
  • Add to Cart