COM448684580 Combi-Blocks, Inc.

1-Chloro-5-fluoro-4-methanesulfonyl-2-nitrobenzene

  • MDL number MFCD27936860
  • Cas Number : 1820740-06-4
  • Molecular Weight : 253.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9495

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9495
Availability details for
Quantity
  • Availability
  • Add to Cart