COM448684585 Combi-Blocks, Inc.

5-(4-Hydroxyphenyl)-3H-1,3,4-oxadiazol-2-one

  • MDL number MFCD24716809
  • Cas Number : 120035-90-7
  • Molecular Weight : 178.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9505

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9505
Availability details for
Quantity
  • Availability
  • Add to Cart