COM448684590 Combi-Blocks, Inc.

5-(2-Methylphenyl)-3H-1,3,4-oxadiazol-2-one

  • MDL number MFCD11046699
  • Cas Number : 119933-32-3
  • Molecular Weight : 176.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9510

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9510
Availability details for
Quantity
  • Availability
  • Add to Cart