COM448684592 Combi-Blocks, Inc.

2-(3-Methoxyphenyl)-1,3,4-oxadiazole

  • MDL number MFCD09755524
  • Cas Number : 5378-30-3
  • Molecular Weight : 176.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9513

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9513
Availability details for
Quantity
  • Availability
  • Add to Cart