COM448684594 Combi-Blocks, Inc.

5-(3-Fluorophenyl)-3H-1,3,4-oxadiazol-2-one

  • MDL number MFCD11046707
  • Cas Number : 1044766-12-2
  • Molecular Weight : 180.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9516

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9516
Availability details for
Quantity
  • Availability
  • Add to Cart