COM448684601 Combi-Blocks, Inc.

5-(2-Bromophenyl)-3H-1,3,4-oxadiazol-2-one

  • MDL number MFCD09746378
  • Cas Number : 1044766-10-0
  • Molecular Weight : 241.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9526

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9526
Availability details for
Quantity
  • Availability
  • Add to Cart