COM448684619 Combi-Blocks, Inc.

2-(Chloromethyl)-5-[3-(trifluoromethyl)phenyl]-1,3,4-oxadiazole

  • MDL number MFCD09816147
  • Cas Number : 944897-58-9
  • Molecular Weight : 262.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9563

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9563
Availability details for
Quantity
  • Availability
  • Add to Cart