COM448684633 Combi-Blocks, Inc.

5-(2,4-Difluorophenyl)-1,3,4-oxadiazol-2-ol

  • MDL number MFCD19673354
  • Cas Number : 1341663-05-5
  • Molecular Weight : 198.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9615

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9615
Availability details for
Quantity
  • Availability
  • Add to Cart