COM448684639 Combi-Blocks, Inc.

3-Bromo-4-chloro-5-nitrobenzoic acid

  • MDL number MFCD24107187
  • Cas Number : 1523065-07-7
  • Molecular Weight : 280.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9623

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9623
Availability details for
Quantity
  • Availability
  • Add to Cart