COM448684647 Combi-Blocks, Inc.

6-Bromo-4-chloro-3H-1,3-benzodiazole

  • MDL number MFCD23704357
  • Cas Number : 1360932-45-1
  • Molecular Weight : 231.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9634

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9634
Availability details for
Quantity
  • Availability
  • Add to Cart