COM448684648 Combi-Blocks, Inc.

6-Bromo-4,5-difluoro-1,3-dihydro-1,3-benzodiazol-2-one

  • MDL number MFCD28101654
  • Cas Number : 1820666-05-4
  • Molecular Weight : 249.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9636

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9636
Availability details for
Quantity
  • Availability
  • Add to Cart