COM448684691 Combi-Blocks, Inc.

Methyl 5-amino-2,4-dichlorobenzoate

  • MDL number MFCD17276520
  • Cas Number : 84228-48-8
  • Molecular Weight : 220.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9712

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9712
Availability details for
Quantity
  • Availability
  • Add to Cart