COM448684705 Combi-Blocks, Inc.

5-Bromo-2-acetamido-4-fluorobenzoic acid

  • MDL number MFCD28143977
  • Cas Number : 1820649-12-4
  • Molecular Weight : 276.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9741

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9741
Availability details for
Quantity
  • Availability
  • Add to Cart