COM448684708 Combi-Blocks, Inc.

4-Bromo-5-chloro-2-nitrobenzamide

  • MDL number MFCD28166416
  • Cas Number : 1629270-60-5
  • Molecular Weight : 279.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9747

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9747
Availability details for
Quantity
  • Availability
  • Add to Cart