COM448684711 Combi-Blocks, Inc.

4-Chloro-2-nitrobenzamide

  • MDL number MFCD01215396
  • Cas Number : 41994-91-6
  • Molecular Weight : 200.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9750

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9750
Availability details for
Quantity
  • Availability
  • Add to Cart