COM448684718 Combi-Blocks, Inc.

Methyl 6-amino-5-bromo-3-chloropyridine-2-carboxylate

  • MDL number MFCD28128287
  • Cas Number : 577691-68-0
  • Molecular Weight : 265.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9776

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9776
Availability details for
Quantity
  • Availability
  • Add to Cart