COM448684727 Combi-Blocks, Inc.

5-[4-(Trifluoromethyl)phenyl]-3H-1,3,4-oxadiazol-2-one

  • MDL number MFCD19673310
  • Cas Number : 202823-22-1
  • Molecular Weight : 230.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9798

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9798
Availability details for
Quantity
  • Availability
  • Add to Cart