COM448684745 Combi-Blocks, Inc.

Dimethyl biphenyl-3,4'-dicarboxylate

  • MDL number MFCD06204223
  • Cas Number : 55676-76-1
  • Molecular Weight : 270.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0006

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0006
Availability details for
Quantity
  • Availability
  • Add to Cart