COM448684746 Combi-Blocks, Inc.

2-Hydroxy-3-(thiophen-2-yl)pyridine

  • MDL number MFCD18311917
  • Cas Number : 30236-48-7
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0014

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0014
Availability details for
Quantity
  • Availability
  • Add to Cart