COM448684752 Combi-Blocks, Inc.

6-(4-Fluorophenyl)-2-hydroxypyridine

  • MDL number MFCD11876187
  • Cas Number : 1111111-04-6
  • Molecular Weight : 189.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0084

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0084
Availability details for
Quantity
  • Availability
  • Add to Cart