COM448684753 Combi-Blocks, Inc.

6-(4-Cyanophenyl)-2-hydroxypyridine

  • MDL number MFCD11876110
  • Cas Number : 1111110-50-9
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0102

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0102
Availability details for
Quantity
  • Availability
  • Add to Cart