COM448684754 Combi-Blocks, Inc.

3-(3-Formylphenyl)-2-hydroxypyridine

  • MDL number MFCD18311961
  • Cas Number : 1261956-40-4
  • Molecular Weight : 199.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0110

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0110
Availability details for
Quantity
  • Availability
  • Add to Cart