COM448684761 Combi-Blocks, Inc.

2-Hydroxy-5-(3-nitrophenyl)pyridine

  • MDL number MFCD11876467
  • Cas Number : 1111106-29-6
  • Molecular Weight : 216.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0389

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0389
Availability details for
Quantity
  • Availability
  • Add to Cart