COM448684763 Combi-Blocks, Inc.

3-(3-Chloro-4-methylphenyl)-2-hydroxypyridine

  • MDL number MFCD18312198
  • Cas Number : 1261938-38-8
  • Molecular Weight : 219.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0458

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0458
Availability details for
Quantity
  • Availability
  • Add to Cart