COM448684764 Combi-Blocks, Inc.

4-(3-Chloro-5-fluorophenyl)-2-hydroxypyridine

  • MDL number MFCD18312233
  • Cas Number : 1261938-64-0
  • Molecular Weight : 223.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0502

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0502
Availability details for
Quantity
  • Availability
  • Add to Cart