COM448684766 Combi-Blocks, Inc.

6-[Benzo(b)thiophen-2-yl]-2-hydroxypyridine

  • MDL number MFCD11876406
  • Cas Number : 1111105-57-7
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0522

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0522
Availability details for
Quantity
  • Availability
  • Add to Cart