COM448684768 Combi-Blocks, Inc.

2-Hydroxy-6-(4-trifluoromethylphenyl)pyridine

  • MDL number MFCD11876114
  • Cas Number : 1111110-54-3
  • Molecular Weight : 239.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0666

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0666
Availability details for
Quantity
  • Availability
  • Add to Cart