COM448684769 Combi-Blocks, Inc.

4-(2,5-Dichlorophenyl)-2-hydroxypyridine

  • MDL number MFCD18312353
  • Cas Number : 1261896-17-6
  • Molecular Weight : 240.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0682

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0682
Availability details for
Quantity
  • Availability
  • Add to Cart