COM448684770 Combi-Blocks, Inc.

6-(2-Benzyloxyphenyl)-2-hydroxypyridine

  • MDL number MFCD11876203
  • Cas Number : 1111111-20-6
  • Molecular Weight : 277.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0876

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0876
Availability details for
Quantity
  • Availability
  • Add to Cart