COM448684771 Combi-Blocks, Inc.

5-(3-BOC-Aminophenyl)-2-hydroxypyridine

  • MDL number MFCD18312548
  • Cas Number : 1261974-19-9
  • Molecular Weight : 286.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0917

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0917
Availability details for
Quantity
  • Availability
  • Add to Cart