COM448684772 Combi-Blocks, Inc.

3-(4-BOC-Aminophenyl)-2-hydroxypyridine

  • MDL number MFCD18312551
  • Cas Number : 1261896-32-5
  • Molecular Weight : 286.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0920

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0920
Availability details for
Quantity
  • Availability
  • Add to Cart