COM448684789 Combi-Blocks, Inc.

4-(2-Hydroxyphenyl)benzoic acid

  • MDL number MFCD06802564
  • Cas Number : 223127-00-2
  • Molecular Weight : 214.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0987

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-0987
Availability details for
Quantity
  • Availability
  • Add to Cart